Sodium hydroxide ≥98%, soluble
Supplier: BeanTown Chemical
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
142245-10KG
142245-2.5KG
142245-500G
BT142245-10KGEA
295.82
USD
BT142245-10KG
BT142245-2.5KG
BT142245-500G
Sodium hydroxide ≥98%, soluble
Sodium hydroxide
CAS: 1310-73-2; EC No: 215-185-5; MDL No: MFCD00003548; RTECS: WB4900000
UN No: UN1823; Haz Class: 8; Packing Group: II
Flakes; Linear Formula: NaOH; MW: 40.00
Melting Point: 318°; Boiling Point: 1390°
Density (g/mL): 2.13
Air Sensitive , Hygroscopic
- RTECS: WB4900000
- Air Sensitive, Hygroscopic
- Flakes
Formula:
NaOH MW: 40 g/mol Boiling Pt: 1390 Melting Pt: 323 Density: 2.13 g/mL |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1823 ADR: 8 II Merck Index: 14,08627 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...