Lithium sulfate, anhydrous ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical

142240-1KG 142240-250G 142240-5KG
BT142240-1KGEA 502.65 USD
BT142240-1KG BT142240-250G BT142240-5KG
Lithium sulfate, anhydrous ≥99.5% (trace metals basis)
Lithium sulphate

CAS: 10377-48-7; EC No: 233-820-4; MDL No: MFCD00011086 -40 Mesh Powder; Linear Formula: Li2SO4; MW: 109.94 Melting Point: 845° Density (g/mL): 2.22 Hygroscopic


  • Hygroscopic
  • -40 Mesh Powder

Formula: Li₂O₄S
Melting Pt: 845 °C
Density: 2.22 g/mL
MDL Number: MFCD00011086
CAS Number: 10377-48-7
EINECS: 233-820-4

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR