1,3-Dibromobenzene ≥97%
Supplier: BeanTown Chemical
142170-100G
142170-25G
142170-500G
BT142170-100GEA
200
USD
BT142170-100G
BT142170-25G
BT142170-500G
1,3-Dibromobenzene ≥97%
1,3-Dibromobenzene
CAS: 108-36-1; EC No: 203-574-2; MDL No: MFCD00000078; RTECS: CZ1790000
UN No: UN3082; Haz Class: 9; Packing Group: III
Liquid; Molecular Formula: C6H4Br2; MW: 235.90
Melting Point: -7°; Boiling Point: 218-219°; Flash point: 93°C (199°F)
Density (g/mL): 1.952; Refractive Index: 1.608
- RTECS: CZ1790000
- Refractive index: 1.608
- Liquid
Formula:
C₆H₄Br₂ Boiling Pt: 218…219 °C Melting Pt: –7 °C Density: 1.952 g/mL Flash Pt: 93 °C (199 °F) |
MDL Number:
MFCD00000078 CAS Number: 108-36-1 EINECS: 203-574-2 ADR: 9,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...