Chroman-4-one ≥98%
Supplier: BeanTown Chemical
Synonyms:
4-Chromanone
142125-100G
142125-25G
142125-5G
BT142125-100GEA
586
USD
BT142125-100G
BT142125-25G
BT142125-5G
Chroman-4-one ≥98%
Chroman-4-one
CAS: 491-37-2; EC No: 207-736-3; MDL No: MFCD00006840; RTECS: DJ2981225
Solid; Molecular Formula: C9H8O2; MW: 148.16
Melting Point: 36-40°; Boiling Point: 127-128°; Flash point: 110°C (230°F)
Refractive Index: 1.575
- RTECS: DJ2981225
- Refractive index: 1.575
- Solid
Formula:
C₉H₈O₂ Boiling Pt: 127…128 °C Melting Pt: 36…40 °C Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00006840 CAS Number: 491-37-2 EINECS: 207-736-3 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...