Ruthenium(III) chloride hydrate ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Ruthenium(III) chloride xhydrate, Ruthenium trichloride hydrate

142070-1G 142070-25G 142070-5G
BT142070-1GEA 57.25 USD
BT142070-1G BT142070-25G BT142070-5G
Ruthenium(III) chloride hydrate ≥99.9%
Ruthenium (III) chloride hydrate

CAS: 14898-67-0; EC No: 233-167-5; MDL No: MFCD00149844; RTECS: VM2650000 UN No: UN3260; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: RuCl3·H2O; MW: 207.43 (anhydrous) Melting Point: 100° (decomposes) Hygroscopic


  • RTECS: VM2650000
  • Hygroscopic
  • Crystalline

Formula: Cl₃Ru·xH₂O
MW: 225.44
Melting Pt: 100 °C (dec.)
Density: 3.11
MDL Number: MFCD00149844
CAS Number: 14898-67-0
EINECS: 233-167-5
UN: 3260
ADR: 8 III

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR