(±)-Mandelic acid ≥99%
Supplier: BeanTown Chemical
Synonyms:
DL-Amygdalic Acid, Mandelic acid, DL-Mandelic acid, 2-Hydroxy-2-phenylacetic acid, (±)-2-Hydroxy-2-phenylacetic acid, DL-2-Hydroxy-2-phenylacetic acid
142050-100G
142050-2.5KG
142050-500G
BT142050-100GEA
27.52
USD
BT142050-100G
BT142050-2.5KG
BT142050-500G
(±)-Mandelic acid ≥99%
(±)-Mandelic acid
CAS: 90-64-2; EC No: 202-007-6; MDL No: MFCD00064250; RTECS: OO6300000
Solid; Linear Formula: C6H5CH(OH)CO2H; Molecular Formula: C8H8O3; MW: 152.15
Melting Point: 119-121°
Light Sensitive
- RTECS: OO6300000
- Light Sensitive
- Solid
Formula:
C₈H₈O₃ MW: 152.15 g/mol Boiling Pt: 322 Melting Pt: 119…121 °C |
MDL Number:
MFCD00064250 CAS Number: 90-64-2 EINECS: 202-007-6 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...