Nitrosium tetrafluoroborate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Nitrosyl tetrafluoroborate, Nitrosonium tetrafluoroborate
142030-1G
142030-25G
142030-5G
BT142030-1GEA
18
USD
BT142030-1G
BT142030-25G
BT142030-5G
Nitrosium tetrafluoroborate ≥98%
Nitrosium tetrafluoroborate
CAS: 14635-75-7; EC No: 238-679-2; MDL No: MFCD00011433
UN No: UN3260; Haz Class: 8; Packing Group: II
Powder; Linear Formula: NOBF4; MW: 116.82
Melting Point: 250°
Density (g/mL): 2.185
Moisture Sensitive
- Moisture Sensitive
- Powder
Formula:
BF₄NO Melting Pt: 250 °C Density: 2.185 g/mL |
MDL Number:
MFCD00011433 CAS Number: 14635-75-7 EINECS: 238-679-2 UN: 3260 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...