1,2,4-Triazole ≥99%
Supplier: BeanTown Chemical
Synonyms:
1,2,4-1H-Triazole, 1H-1,2,4-Triazole
141950-100G
141950-25G
141950-500G
BT141950-100GEA
120
USD
BT141950-100G
BT141950-25G
BT141950-500G
1,2,4-Triazole ≥99%
1,2,4-Triazole
CAS: 288-88-0; EC No: 206-022-9; MDL No: MFCD00005228; RTECS: XZ3806000
Crystalline/Flakes; Molecular Formula: C2H3N3 ; MW: 69.07
Melting Point: 119-121°; Boiling Point: 260°; Flash point: 170°C (338°F)
- RTECS: XZ3806000
- Crystalline/Flakes
Formula:
C₂H₃N₃ Boiling Pt: 260 °C Melting Pt: 119…121 °C Flash Pt: 170 °C (338 °F) |
MDL Number:
MFCD00005228 CAS Number: 288-88-0 EINECS: 206-022-9 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...