Benzyl alcohol ≥99.0% ACS
Supplier: BeanTown Chemical
Synonyms:
Benzenemethanol, Phenylmethanol, Phenylcarbinol
141880-1L
141880-250ML
141880-4L
BT141880-1LEA
125.36
USD
BT141880-1L
BT141880-250ML
BT141880-4L
Benzyl alcohol ≥99.0% ACS
Benzyl alcohol
CAS: 100-51-6; EC No: 202-859-9; MDL No: MFCD00004599; RTECS: DN3150000
Liquid; Molecular Formula: C7H8O ; MW: 108.14
Melting Point: -16°; Boiling Point: 203-205°; Flash point: 96°C (205°F)
Density (g/mL): 1.045; Refractive Index: 1.539
- RTECS: DN3150000
- Refractive index: 1.539
- Liquid
Formula:
C₆H₅CH₂OH Boiling Pt: 203…205 °C Melting Pt: –16 °C Density: 1.045 g/mL Flash Pt: 96 °C (205 °F) |
MDL Number:
MFCD00004599 CAS Number: 100-51-6 EINECS: 202-859-9 |
|
Specification Test Results
Assay | 99.0% min. |
Acetophenone | ≤0.02% |
Benzaldehyde | ≤0.01% |
Color (APHA) | ≤20 |
Residue after ignition | ≤0.005% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...