Tin(II) oxide ≥99.9% (trace metals basis)
Supplier: BeanTown Chemical
141740-100G
141740-25G
141740-500G
BT141740-100GEA
42.57
USD
BT141740-100G
BT141740-25G
BT141740-500G
Tin(II) oxide ≥99.9% (trace metals basis)
Tin (II) oxide
CAS:
21651-19-4; EC No: 244-499-5; MDL No: MFCD00011243; RTECS: XQ3700000
-100 Mesh Powder; Linear Formula: SnO2; MW: 134.69
Melting Point: 1080° (decomposes)
Density (g/mL): 6.45
Air Sensitive
- RTECS: XQ3700000
- Air Sensitive
- -100 Mesh Powder
Formula:
OSn Melting Pt: 1080 °C (dec.) Density: 6.45 g/mL |
MDL Number:
MFCD00011243 CAS Number: 21651-19-4 EINECS: 244-499-5 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...