Lithium manganese(III,IV) oxide ≥99.5%

Supplier: BeanTown Chemical

141725-100G 141725-25G
BT141725-100GEA 271 USD
BT141725-100G BT141725-25G
Lithium manganese(III,IV) oxide ≥99.5%
Lithium manganese(III,IV) oxide

CAS: 12057-17-9; MDL No: MFCD01114233 -325 Mesh Powder; Molecular Formula: LiMn2O4 ; MW: 180.81 Melting Point: 400°


  • -325 Mesh Powder

Formula: LiMn₂O₄
Melting Pt: >400 °C
MDL Number: MFCD01114233
CAS Number: 12057-17-9

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR