2,6-Di-tert-butyl-4-methylpyridine ≥97%

Supplier: BeanTown Chemical

Synonyms: 2,6-Di-Tert-butyl-4-methylpyridine

141720-1G 141720-5G
BT141720-1GEA 35 USD
BT141720-1G BT141720-5G
2,6-Di-tert-butyl-4-methylpyridine ≥97%
2,6-Di-tert-butyl-4-methylpyridine

CAS: 38222-83-2; EC No: 253-834-4; MDL No: MFCD00006305 Powder; Molecular Formula: C14H23N; MW: 205.34 Melting Point: 33-36°; Boiling Point: 233°; Flash point: 84°C (183°F) Refractive Index: 1.476 Air Sensitive, Light Sensitive


  • Refractive index: 1.476
  • Air Sensitive, Light Sensitive
  • Powder

Formula: C₁₄H₂₃N
MW: 205.34 g/mol
Boiling Pt: 233 °C
Melting Pt: 33…36 °C
Flash Pt: 84 °C (183 °F)
MDL Number: MFCD00006305
CAS Number: 38222-83-2
EINECS: 253-834-4

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR