Cobalt ≥99.95% (trace metals basis), slug, Length 3.175 mm, Diameter 3.175 mm
Supplier: BeanTown Chemical
141700-100G
141700-25G
141700-5G
BT141700-100GEA
1076
USD
BT141700-100G
BT141700-25G
BT141700-5G
Cobalt ≥99.95% (trace metals basis), slug, Length 3.175 mm, Diameter 3.175 mm
Cobalt
CAS: 7440-48-4; EC No: 231-158-0; MDL No: MFCD00010935; RTECS: GF8750000
Solid; Molecular Formula: Co; MW: 58.93
Melting Point: 1495°; Boiling Point: 2900°
Density (g/mL): 8.92
- RTECS: GF8750000
- Solid
Formula:
Co Boiling Pt: 2900 °C Melting Pt: 1495 °C Density: 8.92 g/mL |
MDL Number:
MFCD00010935 CAS Number: 7440-48-4 EINECS: 231-158-0 |
Specification Test Results
Assay | 99.95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...