L(-)-Glutathione (reduced form) ≥97%

Supplier: BeanTown Chemical

Synonyms: L-Glutathione Reduced, GSH, (S)-2-Amino-5-(((R)-1-((carboxymethyl)amino)-3-mercapto-1-oxopropan-2-yl)amino)-5-oxopentanoic acid, Glutathione Reduced

141690-100G 141690-25G 141690-5G
BT141690-100GEA 646.17 USD
BT141690-100G BT141690-25G BT141690-5G
L(-)-Glutathione (reduced form) ≥97%
L(-)-Glutathione (reduced form)

CAS: 70-18-8; EC No: 200-725-4; MDL No: MFCD00065939; RTECS: MC0556000 Powder; Molecular Formula: C10H17N3O6S; MW: 307.33 Melting Point: 192-195° (decomposes)


  • RTECS: MC0556000
  • Powder

Formula: C₁₀H₁₇N₃O₆S
Melting Pt: 192…195 °C (dec.)
MDL Number: MFCD00065939
CAS Number: 70-18-8
EINECS: 200-725-4

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR