Discandium trioxide ≥99.995%

Supplier: BeanTown Chemical

Synonyms: Scandium oxide, Scandium (III) oxide

141685-1G 141685-5G
BT141685-1GEA 114 USD
BT141685-1G BT141685-5G
Discandium trioxide ≥99.995%
Discandium trioxide

CAS: 12060-08-1; EC No: 235-042-0; MDL No: MFCD00011223 Powder; Linear Formula: Sc2O3; MW: 137.91 Melting Point: 2485° Density (g/mL): 8.347; Refractive Index: 1.964


  • Refractive index: 1.964
  • Powder

Formula: O₃Sc₂
MW: 137.91
Melting Pt: 1000
Density: 8.347 g/mL
MDL Number: MFCD00011223
CAS Number: 12060-08-1
EINECS: 235-042-0
Merck Index: 14,08392

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR