Ytterbium(III) oxide ≥99.9% (trace rare earth metals basis)
Supplier: BeanTown Chemical
141540-100G
141540-25G
BT141540-100GEA
500
USD
BT141540-100G
BT141540-25G
Ytterbium(III) oxide ≥99.9% (trace rare earth metals basis)
Ytterbium (III) oxide
CAS: 1314-37-0; EC No: 215-234-0; MDL No: MFCD00011290
3-12mm Sintered Lump; Linear Formula: Yb2O3; MW: 394.08
Melting Point: 2227°
Density (g/mL): 9.170
Air Sensitive, Hygroscopic
- Air Sensitive, Hygroscopic
- 3-12mm Sintered Lump
Formula:
O₃Yb₂ Melting Pt: 2227 °C Density: 9.170 g/mL |
MDL Number:
MFCD00011290 CAS Number: 1314-37-0 EINECS: 215-234-0 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...