Citric acid monohydrate ≥99.5%

Supplier: BeanTown Chemical

Synonyms: 2-Hydroxypropane-1,2,3-tricarboxylic acid, Hydroxytricarballylic acid, 2-Hydroxy-1,2,3-propanetricarboxilic acid

141525-100G 141525-1KG 141525-500G
BT141525-100GEA 34.29 USD
BT141525-100G BT141525-1KG BT141525-500G
Citric acid monohydrate ≥99.5%
Citric acid monohydrate

CAS: 5949-29-1; EC No: 201-069-1; MDL No: MFCD00149972; RTECS: GE7810000 Granular; Molecular Formula: C6H8O7·H2O ; MW: 210.14 Density (g/mL): 1.542 Hygroscopic


  • RTECS: GE7810000
  • Hygroscopic
  • Granular

Formula: C₆H₈O₇.H₂O
Density: 1.542 g/mL
MDL Number: MFCD00149972
CAS Number: 5949-29-1
EINECS: 201-069-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3160681f96abd248f56672a96b72cc5f6e9d89a2&vid=c4db52c32c630d2c9876e60fc3e9e70a117ea0d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR