4-tert-Butylphenol ≥99%

Supplier: BeanTown Chemical

141415-2.5KG 141415-500G
BT141415-2.5KGEA 139.86 USD
BT141415-2.5KG BT141415-500G
4-tert-Butylphenol ≥99%
4-tert-Butylphenol

CAS: 98-54-4; EC No: 202-679-0; MDL No: MFCD00002367; RTECS: SJ8925000 UN No: UN3077; Haz Class: 9; Packing Group: III Powder; Molecular Formula: C10H14O; MW: 150.22 Melting Point: 96-101°; Boiling Point: 236-238°; Flash point: 110°C (230°F) Density (g/mL): 0.908


  • RTECS: SJ8925000
  • Powder

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
Boiling Pt: 237
Melting Pt: 96…101 °C
Density: 0.908 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00002367
CAS Number: 98-54-4
EINECS: 202-679-0
UN: 3077
ADR: 8 III
Merck Index: 13,01584

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR