(S)-(-)-4-Fluoro-α-methylbenzylamine ≥99%, ee 99%

Supplier: BeanTown Chemical

Synonyms: (S)-1-(4-Fluorophenyl)ethanamine, (S)-(-)-1-(4-Fluorophenyl)ethylamine

141335-1G 141335-25G 141335-5G
BT141335-1GEA 54 USD
BT141335-1G BT141335-25G BT141335-5G
(S)-(-)-4-Fluoro-α-methylbenzylamine ≥99%, ee 99%
(S)-(-)-4-Fluoro-α-methylbenzylamine

CAS: 66399-30-2; MDL No: MFCD03092999 UN No: UN2735; Haz Class: 8; Packing Group: III Liquid; Molecular Formula: C8H10FN; MW: 139.17 Flash point: 68°C (154°F) Density (g/mL): 1.03 Air Sensitive


  • Air Sensitive
  • Liquid

Formula: C₈H₁₀FN
Density: 1.03 g/mL
Flash Pt: 68 °C (154 °F)
MDL Number: MFCD03092999
CAS Number: 66399-30-2
ADR: 8,III

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR