(S)-(-)-4-Fluoro-α-methylbenzylamine ≥99%, ee 99%
Supplier: BeanTown Chemical
Synonyms:
(S)-1-(4-Fluorophenyl)ethanamine, (S)-(-)-1-(4-Fluorophenyl)ethylamine
141335-1G
141335-25G
141335-5G
BT141335-1GEA
54
USD
BT141335-1G
BT141335-25G
BT141335-5G
(S)-(-)-4-Fluoro-α-methylbenzylamine ≥99%, ee 99%
(S)-(-)-4-Fluoro-α-methylbenzylamine
CAS: 66399-30-2; MDL No: MFCD03092999
UN No: UN2735; Haz Class: 8; Packing Group: III
Liquid; Molecular Formula: C8H10FN; MW: 139.17
Flash point: 68°C (154°F)
Density (g/mL): 1.03
Air Sensitive
- Air Sensitive
- Liquid
Formula:
C₈H₁₀FN Density: 1.03 g/mL Flash Pt: 68 °C (154 °F) |
MDL Number:
MFCD03092999 CAS Number: 66399-30-2 ADR: 8,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...