1,8-Octanediol ≥98%

Supplier: BeanTown Chemical

Synonyms: 1,8-Dihydroxy octane

141185-100G 141185-25G 141185-500G
BT141185-100GEA 105 USD
BT141185-100G BT141185-25G BT141185-500G
1,8-Octanediol ≥98%
1,8-Octanediol

CAS: 629-41-4; EC No: 211-090-8; MDL No: MFCD00002989 Powder; Linear Formula: HO(CH2)8OH; MW: 146.23 Melting Point: 57-61°; Boiling Point: 172°/20 mmHg; Flash point: 120°C (248°F)


  • Powder

Formula: C₈H₁₈O₂
MW: 146.23 g/mol
Boiling Pt: 279
Melting Pt: 57…61 °C
Flash Pt: 120 °C (248 °F)
MDL Number: MFCD00002989
CAS Number: 629-41-4
EINECS: 211-090-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR