2-Fluorophenol ≥98%
Supplier: BeanTown Chemical
Synonyms:
o-Fluorophenol
141170-100G
141170-25G
141170-500G
BT141170-100GEA
176
USD
BT141170-100G
BT141170-25G
BT141170-500G
2-Fluorophenol ≥98%
2-Fluorophenol
CAS: 367-12-4; EC No: 206-681-2; MDL No: MFCD00002155; RTECS: SL4530000
UN No: UN1993 ; Haz Class: 3; Packing Group: III
Liquid; Molecular Formula: C6H5FO; MW: 112.10
Melting Point: 16.1°; Boiling Point: 171-172°/741 mmHg; Flash point: 47°C (117°F)
Density (g/mL): 1.256; Refractive Index: 1.511
- RTECS: SL4530000
- Refractive index: 1.511
- Liquid
Formula:
C₆H₅FO Boiling Pt: 171…172 °C/741 mmHg Melting Pt: 16.1 °C Density: 1.256 g/mL Flash Pt: 47 °C (117 °F) |
MDL Number:
MFCD00002155 CAS Number: 367-12-4 EINECS: 206-681-2 UN: 1993 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...