Ferrocenecarbaldehyde ≥97%
Supplier: BeanTown Chemical
141070-1G
141070-5G
BT141070-1GEA
18.99
USD
BT141070-1G
BT141070-5G
Ferrocenecarbaldehyde ≥97%
Ferrocenecarbaldehyde
CAS: 12093-10-6; EC No: 235-158-1; MDL No: MFCD00001429
Crystalline; Molecular Formula: C11H10FeO; MW: 214.05
Melting Point: 118-120°
Air Sensitive
- Air Sensitive
- Crystalline
Formula:
C₁₁H₁₀FeO Melting Pt: 118…120 °C |
MDL Number:
MFCD00001429 CAS Number: 12093-10-6 EINECS: 235-158-1 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...