Pyridinium tribromide ≥90%, Technical Grade
Supplier: BeanTown Chemical
Synonyms:
Pyridinium bromide perbromide, Monopyridin-1-ium tribromide, Pyridine hydrobromide perbromide
140975-100G
140975-25G
140975-500G
BT140975-100GEA
94.79
USD
BT140975-100G
BT140975-25G
BT140975-500G
Pyridinium tribromide ≥90%, Technical Grade
Pyridinium tribromide
CAS: 39416-48-3; EC No: 254-446-8; MDL No: MFCD00013223
UN No: UN3261; Haz Class: 8; Packing Group: III
Crystalline; Molecular Formula: C5H5N·HBr·Br2; MW: 319.82
Melting Point: 128-136°
- Crystalline
Formula:
C₅H₆Br₃N MW: 319.82 g/mol Melting Pt: 128…136 °C |
MDL Number:
MFCD00013223 CAS Number: 39416-48-3 EINECS: 254-446-8 UN: 3261 ADR: 8 III |
Specification Test Results
Assay | 90% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...