Tantalum carbide ≥99.5%
Supplier: BeanTown Chemical
Synonyms:
Tantalum(IV) carbide
140895-10G
140895-250G
140895-50G
BT140895-10GEA
58
USD
BT140895-10G
BT140895-250G
BT140895-50G
Tantalum carbide ≥99.5%
Tantalum carbide
CAS: 12070-06-3; EC No: 235-118-3; MDL No: MFCD00011255
UN No: UN3178; Haz Class: 4.1; Packing Group: III
-325 Mesh Powder; Linear Formula: TaC; MW: 192.96
Melting Point: 3880°
- -325 Mesh Powder
Formula:
CTa Melting Pt: 3880 °C |
MDL Number:
MFCD00011255 CAS Number: 12070-06-3 EINECS: 235-118-3 UN: 3178 ADR: 4.1,III |
Specification Test Results
Assay | 99.5% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...