Tantalum carbide ≥99.5%

Supplier: BeanTown Chemical

Synonyms: Tantalum(IV) carbide

140895-10G 140895-250G 140895-50G
BT140895-10GEA 58 USD
BT140895-10G BT140895-250G BT140895-50G
Tantalum carbide ≥99.5%
Tantalum carbide

CAS: 12070-06-3; EC No: 235-118-3; MDL No: MFCD00011255 UN No: UN3178; Haz Class: 4.1; Packing Group: III -325 Mesh Powder; Linear Formula: TaC; MW: 192.96 Melting Point: 3880°


  • -325 Mesh Powder

Formula: CTa
Melting Pt: 3880 °C
MDL Number: MFCD00011255
CAS Number: 12070-06-3
EINECS: 235-118-3
UN: 3178
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR