Methylphosphonic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
Methanephosphonic acid
140835-1G
140835-25G
140835-5G
BT140835-1GEA
21
USD
BT140835-1G
BT140835-25G
BT140835-5G
Methylphosphonic acid ≥98%
Methylphosphonic acid
CAS: 993-13-5; EC No: 213-607-2; MDL No: MFCD00002137
UN No: UN3261; Haz Class: 8; Packing Group: II
Solid; Linear Formula: CH3P(O)(OH)2; Molecular Formula: CH5O3P; MW: 96.03
Melting Point: 105-107°
- Solid
Formula:
CH₅O₃P Melting Pt: 105…107 °C |
MDL Number:
MFCD00002137 CAS Number: 993-13-5 EINECS: 213-607-2 UN: 3261 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...