Isosorbide dimethyl ether ≥98%

Supplier: BeanTown Chemical

Synonyms: (3R,3aR,6S,6aR)-3,6-Dimethoxyhexahydrofuro[3,2-b]furan

140735-100G 140735-25G
BT140735-100GEA 180 USD
BT140735-100G BT140735-25G
Isosorbide dimethyl ether ≥98%
Isosorbide dimethyl ether

CAS: 5306-85-4; EC No: 226-159-8; MDL No: MFCD00064828 Liquid; Molecular Formula: C8H14O4; MW: 174.20 Boiling Point: 93-95°/0.1 mmHg; Flash point: 108°C (226°F) Density (g/mL): 1.15; Refractive Index: 1.462


  • Refractive index: 1.462
  • Liquid

Formula: C₈H₁₄O₄
Boiling Pt: 93…95 °C/0.1 mmHg
Density: 1.15 g/mL
Flash Pt: 108 °C (226 °F)
MDL Number: MFCD00064828
CAS Number: 5306-85-4
EINECS: 226-159-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR