4-Phenylbutyric acid ≥99%

Supplier: BeanTown Chemical

Synonyms: 4-Phenylbutanoic acid

140580-100G 140580-25G 140580-500G
BT140580-100GEA 184 USD
BT140580-100G BT140580-25G BT140580-500G
4-Phenylbutyric acid ≥99%
4-Phenylbutyric acid

CAS: 1821-12-1; EC No: 217-341-8; MDL No: MFCD00004403 Crystalline; Linear Formula: C6H5(CH2)3CO2H; Molecular Formula: C10H12O2; MW: 164.20 Melting Point: 49-51°; Boiling Point: 165°/10 mmHg; Flash point: 110°C (230°F)


  • Crystalline

Formula: C₁₀H₁₂O₂
Boiling Pt: 165 °C/10 mmHg
Melting Pt: 49…51 °C
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00004403
CAS Number: 1821-12-1
EINECS: 217-341-8

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR