Lithium fluoride ≥99.99%
Supplier: BeanTown Chemical
140440-10G
140440-50G
BT140440-10GEA
85.27
USD
BT140440-10G
BT140440-50G
Lithium fluoride ≥99.99%
Lithium fluoride
CAS: 7789-24-4; EC No: 232-152-0; MDL No: MFCD00011090; RTECS: OJ6125000
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: LiF; MW: 25.94
Melting Point: 845°; Boiling Point: 1673°
Density (g/mL): 2.64
- RTECS: OJ6125000
- Powder
Formula:
FLi Boiling Pt: 1673 °C Melting Pt: 845 °C Density: 2.64 g/mL |
MDL Number:
MFCD00011090 CAS Number: 7789-24-4 EINECS: 232-152-0 UN: 3288 ADR: 6.1,III |
Specification Test Results
Assay | 99.99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...