N-Phthaloylglycine ≥98%

Supplier: BeanTown Chemical

Synonyms: (1,3-Dihydro-1,3-dioxo-2H-isoindol-2-yl)acetic acid

140415-1KG 140415-250G 140415-50G
BT140415-1KGEA 640 USD
BT140415-1KG BT140415-250G BT140415-50G
N-Phthaloylglycine ≥98%
N-Phthaloylglycine

CAS: 4702-13-0; EC No: 225-177-3; MDL No: MFCD00005900; RTECS: NR3439000 Crystalline; Molecular Formula: C10H7NO4 ; MW: 205.17 Melting Point: 193-196°


  • RTECS: NR3439000
  • Crystalline

Formula: C₁₀H₇NO₄
MW: 205.17 g/mol
Melting Pt: 193…196 °C
MDL Number: MFCD00005900
CAS Number: 4702-13-0
EINECS: 225-177-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR