Aluminium oxide, regular desiccant
Supplier: BeanTown Chemical
Synonyms:
Aluminium(III) oxide, Alumina C, Aluminum oxide
140385-100G
140385-2KG
140385-500G
BT140385-100GEA
27.98
USD
BT140385-100G
BT140385-2KG
BT140385-500G
Aluminium oxide, regular desiccant
Aluminium oxide
CAS: 1344-28-1; EC No: 215-691-6; MDL No: MFCD00003424
3.2mm Spheres; Molecular Formula: Al2O3; MW: 101.96
Melting Point: 2040°
- 3.2mm Spheres
Formula:
Al₂O₃ Melting Pt: 2040 °C |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...