Strontium carbonate ≥97%

Supplier: BeanTown Chemical

140345-1KG 140345-250G 140345-5KG
BT140345-1KGEA 50 USD
BT140345-1KG BT140345-250G BT140345-5KG
Strontium carbonate ≥97%
Strontium carbonate

CAS: 1633-05-2; EC No: 216-643-7; MDL No: MFCD00011250 Powder; Linear Formula: SrCO3; MW: 147.63 Melting Point: 1100° (decomposes) Density (g/mL): 3.70


  • Powder

Formula: CO₃Sr
Melting Pt: 1100 °C (dec.)
Density: 3.70 g/mL
MDL Number: MFCD00011250
CAS Number: 1633-05-2
EINECS: 216-643-7

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR