Rhodizonic acid disodium salt ≥98%

Supplier: BeanTown Chemical

Synonyms: Sodium rhodizonate

140340-100G 140340-25G 140340-5G
BT140340-100GEA 579.38 USD
BT140340-100G BT140340-25G BT140340-5G
Rhodizonic acid disodium salt ≥98%
Rhodizonic acid disodium salt

CAS: 523-21-7; EC No: 208-340-3; MDL No: MFCD00001650 Powder; Molecular Formula: C6Na2O6; MW: 214.04 Melting Point: 300°


  • Powder

Formula: C₆Na₂O₆
Melting Pt: >300 °C
MDL Number: MFCD00001650
CAS Number: 523-21-7
EINECS: 208-340-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR