N-Acetyl-DL-methionine ≥99%
Supplier: BeanTown Chemical
Synonyms:
Ac-DL-Met-OH
140290-1KG
140290-250G
140290-50G
BT140290-1KGEA
136
USD
BT140290-1KG
BT140290-250G
BT140290-50G
N-Acetyl-DL-methionine ≥99%
N-Acetyl-DL-methionine
CAS: 1115-47-5; EC No: 214-224-3; MDL No: MFCD00008681; RTECS: PD0500000
Powder; Linear Formula: CH3SCH2CH2CH(NHCOCH3)COOH; Molecular Formula: C7H13NO3S; MW: 191.25
Melting Point: 117-119°
- RTECS: PD0500000
- Powder
Formula:
C₇H₁₃NO₃S Melting Pt: 117…119 °C |
MDL Number:
MFCD00008681 CAS Number: 1115-47-5 EINECS: 214-224-3 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...