1,5-Dibromopentane ≥98%

Supplier: BeanTown Chemical

140265-100G 140265-2.5KG 140265-500G
BT140265-100GEA 40 USD
BT140265-100G BT140265-2.5KG BT140265-500G
1,5-Dibromopentane ≥98%
1,5-Dibromopentane

CAS: 111-24-0; EC No: 203-849-7; MDL No: MFCD00000268; RTECS: SA0320000 Liquid; Linear Formula: Br(CH2)5Br; Molecular Formula: C5H10Br2 ; MW: 229.94 Melting Point: -34°; Boiling Point: 110°/15 mmHg; Flash point: 79°C (174°F) Density (g/mL): 1.688; Refractive Index: 1.512


  • RTECS: SA0320000
  • Refractive index: 1.512
  • Liquid

Formula: BrH₂C(CH₂)₃CH₂Br
MW: 229.94 g/mol
Boiling Pt: 224
Melting Pt: -39
Density: 1.688 g/mL
Flash Pt: 79 °C (174 °F)
MDL Number: MFCD00000268
CAS Number: 111-24-0
EINECS: 203-849-7

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR