Nitro blue tetrazolium chloride (NBT) ≥90%, Technical Grade
Supplier: BeanTown Chemical
140245-500MG
BT140245-500MGEA
95.85
USD
BT140245-500MG
Nitro blue tetrazolium chloride (NBT) ≥90%, Technical Grade
Nitro blue tetrazolium chloride (NBT)
CAS: 298-83-9; EC No: 206-067-4; MDL No: MFCD00150013; RTECS: XF8045000
Solid; Molecular Formula: C40H30N10O6·2Cl; MW: 817.65
Melting Point: 189°
Air Sensitive, Light Sensitive
- RTECS: XF8045000
- Air Sensitive, Light Sensitive
- Solid
Formula:
C₄₀H₃₀Cl₂N₁₀O₆ Melting Pt: 189 °C |
MDL Number:
MFCD00012159 CAS Number: 298-83-9 EINECS: 206-067-4 |
Specification Test Results
Assay | 90% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...