Iron ≥99.95% (trace metals basis), rods Ø 7.62 mm

Supplier: BeanTown Chemical

140235-190MM
BT140235-190MMEA 118 USD
BT140235-190MM
Iron ≥99.95% (trace metals basis), rods Ø 7.62 mm
Iron

CAS: 7439-89-6; EC No: 231-086-4; MDL No: MFCD00010999 Solid; Molecular Formula: Fe; MW: 55.85 Melting Point: 1535°; Boiling Point: 2750° Density (g/mL): 7.86


  • Solid

Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000
Melting Pt: 1535
Density: 7.86 g/mL
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-086-4
Merck Index: 13,05109

Order Now

Specification Test Results

Assay 99.95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR