Lithium hydroxide, anhydrous ≥98%
Supplier: BeanTown Chemical
140225-100G
140225-25G
140225-500G
BT140225-100GEA
99.08
USD
BT140225-100G
BT140225-25G
BT140225-500G
Lithium hydroxide, anhydrous ≥98%
Lithium hydroxide
CAS: 1310-65-2; EC No: 215-183-4; MDL No: MFCD00011095; RTECS: OJ6307070
UN No: UN2680; Haz Class: 8; Packing Group: II
-4+14 Mesh Powder; Linear Formula: LiOH; MW: 23.95
Melting Point: 470° (decomposes)
Air Sensitive , Hygroscopic
- RTECS: OJ6307070
- Air Sensitive, Hygroscopic
- -4+14 Mesh Powder
Formula:
LiOH Melting Pt: 470 °C (dec.) |
MDL Number:
MFCD00011095 CAS Number: 1310-65-2 EINECS: 215-183-4 UN: 2680 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...