Oxalic acid, anhydrous ≥98%
Supplier: BeanTown Chemical
Synonyms:
Ethanedioic acid
140220-250G
140220-50G
BT140220-250GEA
58.77
USD
BT140220-250G
BT140220-50G
Oxalic acid, anhydrous ≥98%
Oxalic acid
CAS: 144-62-7; EC No: 205-634-3; MDL No: MFCD00002573; RTECS: RO2450000
UN No: UN3261; Haz Class: 8; Packing Group: III
Crystalline; Linear Formula: HOOCCOOH; Molecular Formula: C2H2O4; MW: 90.04
Melting Point: 190° (decomposes); Boiling Point: 157° (sublimes)
Density (g/mL): 1.9
- RTECS: RO2450000
- Crystalline
Formula:
C₂H₂O₄ MW: 90.04 g/mol Boiling Pt: 365 (subl.) Melting Pt: 190 °C (dec.) Density: 1.9 g/mL |
MDL Number:
MFCD00002573 CAS Number: 144-62-7 EINECS: 205-634-3 UN: 3261 ADR: 8 III Merck Index: 13,06980 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...