2-Amino-5-fluoropyridine ≥97%
Supplier: BeanTown Chemical
140165-1G
140165-250MG
140165-5G
BT140165-1GEA
125
USD
BT140165-1G
BT140165-250MG
BT140165-5G
2-Amino-5-fluoropyridine ≥97%
2-Amino-5-fluoropyridine
CAS: 21717-96-4; MDL No: MFCD01861120
UN No: UN2811; Haz Class: 6.1; Packing Group: III
Crystalline; Molecular Formula: C5H5FN2 ; MW: 112.11
Melting Point: 93-97°
- Crystalline
Formula:
C₅H₅FN₂ MW: 112.11 Melting Pt: 93…97 °C |
MDL Number:
MFCD01861120 CAS Number: 21717-96-4 UN: 2811 ADR: 6.1 III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...