Scandium(III) nitrate hydate ≥99.99%

Supplier: BeanTown Chemical
Warning

Synonyms: Trinitric acid scandium salt hydrate, Scandium(3+) trinitrate hydrate, Scandium trinitrate hydrate

140160-1G 140160-5G
BT140160-1GEA 145.05 USD
BT140160-1G BT140160-5G
Scandium(III) nitrate hydate ≥99.99%
Scandium (III) nitrate

CAS: 13465-60-6; EC No: 236-701-5; MDL No: MFCD00011220 UN No: UN1477; Haz Class: 5.1; Packing Group: III Crystalline Aggregates; Linear Formula: Sc(NO3)3·xH2O; MW: 230.97 (anhydrous) Hygroscopic


  • Crystalline Aggregates
  • Hygroscopic

Formula: Sc(NO₃)₃·xH₂O
MW: 230.97
MDL Number: MFCD00011220
CAS Number: 13465-60-6
EINECS: 236-701-5
UN: 1477
ADR: 5.1 III

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR