Uracil ≥99%

Supplier: BeanTown Chemical

Synonyms: Pyrimidine-2,4(1H,3H)-dione

140100-1KG 140100-250G 140100-50G
BT140100-1KGEA 354 USD
BT140100-1KG BT140100-250G BT140100-50G
Uracil ≥99%
Uracil

CAS: 66-22-8; EC No: 200-621-9; MDL No: MFCD00006016; RTECS: YQ8650000 Powder; Molecular Formula: C4H4N2O2; MW: 112.09 Melting Point: 300°


  • RTECS: YQ8650000
  • Powder

Formula: C₄H₄N₂O₂
Melting Pt: >300 °C
MDL Number: MFCD00006016
CAS Number: 66-22-8
EINECS: 200-621-9

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR