1-(Chloromethoxy)propane
Supplier: BeanTown Chemical
139990-1G
139990-5G
BT139990-1GEA
318
USD
BT139990-1G
BT139990-5G
1-(Chloromethoxy)propane
1-(Chloromethoxy)propane
CAS: 3587-57-3
Solid; Linear Formula: CH3CH2CH2OCH2Cl; Molecular Formula: C4H9ClO; MW: 108.57
Boiling Point: 94°; Flash point: 25°C (77°F)
Density (g/mL): 0.967
- Solid
Formula:
C₄H₉ClO MW: 108.565 g/mol Boiling Pt: 94 °C Density: 0.967 g/mL Flash Pt: 25 °C (77 °F) |
CAS Number:
3587-57-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...