Benzenesulfonic acid ≥95%

Supplier: BeanTown Chemical

139890-100G 139890-2.5KG 139890-500G
BT139890-100GEA 26.21 USD
BT139890-100G BT139890-2.5KG BT139890-500G
Benzenesulfonic acid ≥95%
Benzenesulphonic acid

CAS: 98-11-3; EC No: 202-638-7; MDL No: MFCD00011689; RTECS: DB4200000 UN No: UN2585; Haz Class: 8; Packing Group: III Solid; Linear Formula: C6H5SO3H; MW: 158.18 Hygroscopic


  • RTECS: DB4200000
  • Hygroscopic
  • Solid

Formula: C₆H₆O₃S
MW: 158.18 g/mol
Boiling Pt: 319
Melting Pt: 50
MDL Number: MFCD00011689
CAS Number: 98-11-3
EINECS: 202-638-7
UN: 2585
ADR: 8 III
Merck Index: 13,01070

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR