Benzenesulfonic acid ≥95%
Supplier: BeanTown Chemical
139890-100G
139890-2.5KG
139890-500G
BT139890-100GEA
26.21
USD
BT139890-100G
BT139890-2.5KG
BT139890-500G
Benzenesulfonic acid ≥95%
Benzenesulphonic acid
CAS: 98-11-3; EC No: 202-638-7; MDL No: MFCD00011689; RTECS: DB4200000
UN No: UN2585; Haz Class: 8; Packing Group: III
Solid; Linear Formula: C6H5SO3H; MW: 158.18
Hygroscopic
- RTECS: DB4200000
- Hygroscopic
- Solid
Formula:
C₆H₆O₃S MW: 158.18 g/mol Boiling Pt: 319 Melting Pt: 50 |
MDL Number:
MFCD00011689 CAS Number: 98-11-3 EINECS: 202-638-7 UN: 2585 ADR: 8 III Merck Index: 13,01070 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...