Polyethylene oxide, M.W. ∼1000000
Supplier: BeanTown Chemical
Synonyms:
PEO
139870-100G
139870-25G
139870-500G
BT139870-100GEA
78
USD
BT139870-100G
BT139870-25G
BT139870-500G
Polyethylene oxide, M.W. ∼1000000
Polyethylene oxide
CAS: 25322-68-3; EC No: 500-038-2; MDL No: MFCD00147971
Powder; Linear Formula: (-CH2CH2O-)n; MW ≈1,000,000
Boiling Point:
- Powder
Formula:
(-CH₂CH₂O-)n Boiling Pt: >200 °C |
MDL Number:
MFCD00147970 CAS Number: 25322-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...