Molybdic acid ≥85.0% ACS

Supplier: BeanTown Chemical

139840-2KG 139840-500G
BT139840-2KGEA 487 USD
BT139840-2KG BT139840-500G
Molybdic acid ≥85.0% ACS
Molybdic acid

CAS: 7782-91-4; EC No: 231-970-5; MDL No: MFCD00036287 Crystalline/Powder; Molecular Formula: H2MoO4; MW: 161.95 Melting Point: 300° Density (g/mL): 3.1


  • Crystalline/Powder

Formula: H₂MoO₄
MW: 161.95 g/mol
Boiling Pt: 300
Melting Pt: 300
Density: 3.1 g/mL
MDL Number: MFCD00036287
CAS Number: 7782-91-4
EINECS: 231-970-5
Merck Index: 13,06262

Order Now

Specification Test Results

Assay, as MO3 85.0% min.
Insoluble in dilute ammonium hydroxide ≤0.01%
Chloride ≤0.002%
Arsenate, phosphate, and silicate (as SiO2) ≤0.001%
Phosphate (PO4) ≤5ppm
Sulfate (SO4) ≤0.2%
Heavy metals (as Pb) ≤0.003%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR