1-Naphthaldehyde ≥97%
Supplier: BeanTown Chemical
Synonyms:
α-Naphthaldehyde
139665-1KG
139665-250G
139665-50G
BT139665-1KGEA
455
USD
BT139665-1KG
BT139665-250G
BT139665-50G
1-Naphthaldehyde ≥97%
1-Naphthaldehyde
CAS: 66-77-3; EC No: 200-633-4; MDL No: MFCD00004003; RTECS: QJ0190000
Liquid; Molecular Formula: C11H8O; MW: 156.18
Melting Point: 1-2°; Boiling Point: 160-161°/15 mmHg; Flash point: 110°C (230°F)
Density (g/mL): 1.15; Refractive Index: 1.652
Air Sensitive
- RTECS: QJ0190000
- Refractive index: 1.652
- Air Sensitive
- Liquid
Formula:
C₁₁H₈O Boiling Pt: 160…161 °C/15 mmHg Melting Pt: 1…2 °C Density: 1.15 g/mL Flash Pt: 110 °C (230 °F) |
MDL Number:
MFCD00004003 CAS Number: 66-77-3 EINECS: 200-633-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...