Ammonium persulfate (APS) ≥98.0% ACS

Supplier: BeanTown Chemical

Synonyms: Peroxidisulfuric acid diammonium salt, Ammonium persulfate, Peroxidisulphuric acid diammonium salt, Ammonium persulphate

139605-1KG 139605-250G
BT139605-1KGEA 110.52 USD
BT139605-1KG BT139605-250G
Ammonium persulfate (APS) ≥98.0% ACS
Ammonium peroxodisulphate (APS)

CAS: 7727-54-0; EC No: 231-786-5; MDL No: MFCD00003390; RTECS: SE0350000 UN No: UN1444; Haz Class: 5.1; Packing Group: III Lump; Linear Formula: (NH4)2S2O8; MW: 228.20 Moisture Sensitive


  • RTECS: SE0350000
  • Moisture Sensitive
  • Lump

Formula: (NH₄)₂S₂O₈
MW: 228.2 g/mol
Melting Pt: 120
MDL Number: MFCD00003390
CAS Number: 7727-54-0
EINECS: 231-786-5
UN: 1444
ADR: 5.1 III
Merck Index: 13,00545
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f82a373f21d34e63dc94c0598f3f33ba6bbfdb2b&vid=2b6d196dbf07ce4dd528ac7b4f8456678accd3ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR